ContactUsLinkComponent
InfinityLab Poroshell 120 EC-C18, 2.1 x 100 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 %. En dcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Por 1 * 1 Pce
N° de catalogue HEWL695775-902T
Fournisseur:  Agilent
InfinityLab Poroshell 120 EC-C18, 2.1 x 100 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 %. En dcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Por 1 * 1 Pce
N° de catalogue HEWL695775-902T
Fournisseur:  Agilent
Frequently Bought Together