ContactUsLinkComponent
InfinityLab Poroshell 120 EC-C18, 4.6 x 150 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Carbon Load: 10 %. En dcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Por 1 * 1 Pce
  HEWL693975-902T
 :  Agilent
InfinityLab Poroshell 120 EC-C18, 4.6 x 150 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Carbon Load: 10 %. En dcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Por 1 * 1 Pce
  HEWL693975-902T
 :  Agilent
Frequently Bought Together