InfinityLab Poroshell 120 EC-CN, 2.1,150 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 3.5 %. End capped: Double. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab P 1 * 1 Pce
N° de catalogue HEWL693775-905T
Fournisseur: Agilent
Frequently Bought Together