InfinityLab Poroshell 120 PFP, 2.1 x 150 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5.1 %. End capped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poro 1 * 1 Pce
N° de catalogue HEWL693775-408T
Fournisseur: Agilent
Frequently Bought Together