InfinityLab Poroshell 120 PFP, 3.0 x 150 mm, 1.9 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 5.1 %. E ndcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Poro 1 * 1 Pce
N° de catalogue HEWL693675-308
Fournisseur: Agilent
Frequently Bought Together