InfinityLab Poroshell 120 EC-C8, 2.1, 30 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 5 %. Endca pped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Porosh 1 * 1 Pce
HEWL691775-906T
: Agilent
Frequently Bought Together