InfinityLab Poroshell 120 EC-C18, 2.1, 30 mm, 2.7 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 10 %. End capped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poro 1 * 1 Pce
HEWL691775-902T
: Agilent
Frequently Bought Together