InfinityLab Poroshell 120 EC-C18, 4.6, 250 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Carbon Load: 10 %. End capped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poro 1 * 1 Pce
N° de catalogue HEWL690975-902T
Fournisseur: Agilent
Frequently Bought Together