Specifications:- Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C . LC Column Phase: SB-Aq. LC Platform: HPLC. Length: 50 mm. Particle Size: 4 um. Pore Size: 120 um. 1 * 1 Pce
N° de catalogue HEWL689970-914
Fournisseur: Agilent
Frequently Bought Together