ContactUsLinkComponent
Specifications:- Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion UHPLC. Length: 50 mm. Particle Size: 1.9 1 * 1 Pce
  HEWL689675-914
 :  Agilent
undefined
Specifications:- Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion UHPLC. Length: 50 mm. Particle Size: 1.9 1 * 1 Pce
  HEWL689675-914
 :  Agilent
Frequently Bought Together