InfinityLab Poroshell 120 SB-C8, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 4.5 %. Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Por 1 * 1 Pce
N° de catalogue HEWL683975-306T
InfinityLab Poroshell 120 SB-C8, 3.0 x 150 mm, 2.7 um, solvent saver LC column, with column ID. Specifications:- Carbon Load: 4.5 %. Endcapped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 3 mm. LC Column Brand: InfinityLab Por 1 * 1 Pce
N° de catalogue HEWL683975-306T
Frequently Bought Together