Specifications:- Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 80 C . LC Column Phase: SB-Aq. LC Platform: UHPLC Low Dispersion UHPLC. Length: 150 mm. Particle Size: 1. 1 * 1 Pce
N° de catalogue HEWL683675-914
Fournisseur: Agilent
Frequently Bought Together