ContactUsLinkComponent
InfinityLab Poroshell 120 SB-C18, 4.6, 30 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Carbon Load: 7.5 %. End capped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
N° de catalogue HEWL681975-902T
Fournisseur:  Agilent
InfinityLab Poroshell 120 SB-C18, 4.6, 30 mm, 2.7 um, analytical LC column, with column ID. Specifications:- Carbon Load: 7.5 %. End capped: No. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poros 1 * 1 Pce
N° de catalogue HEWL681975-902T
Fournisseur:  Agilent
Frequently Bought Together