PL HFIPgel, 4.6 x 250 mm, 9 um, HPLC column. Specifications:- Guard Column: No. Inner Diameter (ID): 4.6 mm. LC Column Brand: PLgel. LC Column Maximum Temperature: 50 C (HFIP) . Length: 250 mm. Molecular Weight Lower Limit: 200 Da. Molecular Weight 0 1 * 1 Pce
N° de catalogue HEWLPL1514-5900HFI
Fournisseur: Agilent
Frequently Bought Together