Specifications:- Diameter: 2.7 um. Magnetic Particle Iron Content: ~ 20 %. Streptavidin Bead Concentration: 5 % w/v. Streptavidin Bi nding Capacity Biotin: 3,000 pmoles/mg bead. Streptavidin Binding Capacity IgG: 6.5 ug/mg bead. Volume: 400 mL. 1 * 400 mL
HEWLPL6827-1006
: Agilent
Frequently Bought Together