ContactUsLinkComponent
Specifications:- Capillary Material: PTFE. Inner Diameter (ID): 1.5 mm. LC Use With Instrument: G7158B. Outer Diameter (OD): 3.5 mm. Swage Status Fitting Left End: Pre-swaged. Swage Status Fitting Right End: Pre-swaged. 1 * 1 Pce
N° de catalogue HEWL5500-1429
Fournisseur:  Agilent
Specifications:- Capillary Material: PTFE. Inner Diameter (ID): 1.5 mm. LC Use With Instrument: G7158B. Outer Diameter (OD): 3.5 mm. Swage Status Fitting Left End: Pre-swaged. Swage Status Fitting Right End: Pre-swaged. 1 * 1 Pce
N° de catalogue HEWL5500-1429
Fournisseur:  Agilent
Frequently Bought Together