Specifications:- Capillary Material: MP35N. Fitting Type Left End: SI. Fitting Type Right End: SI. Inner Diameter (ID): 0.25 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC System Compatibility: 1290 infinity II Bio 1 * 1 Pce
Specifications:- Capillary Material: MP35N. Fitting Type Left End: SI. Fitting Type Right End: SI. Inner Diameter (ID): 0.25 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC System Compatibility: 1290 infinity II Bio 1 * 1 Pce