ContactUsLinkComponent
Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: SI. Fitting Type Right End : SX. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC Sy 1 * 1 Pce
N° de catalogue HEWL5004-0040
Fournisseur:  Agilent
Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: SI. Fitting Type Right End : SX. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC Sy 1 * 1 Pce
N° de catalogue HEWL5004-0040
Fournisseur:  Agilent
Frequently Bought Together