ContactUsLinkComponent
Specifications:- Chemical Compatibility: Organic solvents. Flared: Yes. Inner Diameter (ID): 0.76 mm. Number of Stops: 2. Spectrosco py Instrument Compatibility: 7500 7700 7800 7850 7900 8800 8900 All Agilent ICP-MS systems. Spectroscopy Tubing Mater 1 * 12 Pce
N° de catalogue HEWL5005-0026
Fournisseur:  Agilent
undefined
Specifications:- Chemical Compatibility: Organic solvents. Flared: Yes. Inner Diameter (ID): 0.76 mm. Number of Stops: 2. Spectrosco py Instrument Compatibility: 7500 7700 7800 7850 7900 8800 8900 All Agilent ICP-MS systems. Spectroscopy Tubing Mater 1 * 12 Pce
N° de catalogue HEWL5005-0026
Fournisseur:  Agilent
Frequently Bought Together