Masterflex® Port Disc Opposing Barb ¼" Int.Ø in Engage 8402 Cleanroom Manufactured; 10 pack
N° de catalogue MFLX40331-13
Fournisseur: Avantor Fluid Handling
À propos de cet article
Designed for sample port and dip tube applications.
- Barb sizes range from 1/8" to 1/2"
- 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.
- Taille de l'ardillon/Tube Ø int. (port 1):¼"
- Type de connection:Port Disc to Barb
- Flow pattern:Straight
- Désignation:Port disc opposing barb ¼" int.ø in engage 8402 cleanroom manufactured; 10 pack
- Environmentally Preferable:
- Regulatory approvals:FDA;USP
- Sterilisation:Gamma irradiation
- Cdt:10
- Fitting material:Engage 8402
Frequently Bought Together