Masterflex® L/S® Easy-Load® Pump Head Replacement Main Body Assembly for 07514-12 and -32
Catalogus # MFLX07514-17
Leverancier: Avantor Fluid Handling
1 year warranty
Over dit artikel
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Désignation:L/S® Easy-Load® Pump Head Replacement Main Body Assembly for 07514-12 and -32
- Environmentally Preferable:
- Open-head sensor:No
- Tubing sizes accepted:L/S® 15, L/S 24
- Garantie:1 year
- Cdt:1
- Matériau du boîtier:Polyarylamide (PARA)
Frequently Bought Together