Masterflex® Fitting, PVDF, Elbow, Hose Barb Union, 3/4" ID"; 10/PK
N° de catalogue MFLX40614-88
Fournisseur: Avantor Fluid Handling
À propos de cet article
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- Taille de l'ardillon/Tube Ø int. (port 1):3/4"
- Taille de l'ardillon/Tube Ø int. (port 2):3/4"
- Type de connection:Barbed Union
- Flow pattern:Elbow
- Désignation:Fitting, PVDF, Elbow, Hose Barb Union, 3/4" ID"; 10/PK
- Environmentally Preferable:
- Orifice dimensions:14,29 mm (0,5625")
- Sterilisation:Autoclave or ETO
- Cdt:10
- Fitting material:PVDF
Frequently Bought Together