Masterflex® Fitting, PVDF, Elbow, Hose Barb Union, 3/32" ID; 10/PK
Catalog # MFLX40614-81
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- Taille de l'ardillon/Tube Ø int. (port 1):3/32"
- Taille de l'ardillon/Tube Ø int. (port 2):3/32"
- Type de connection:Barbed Union
- Flow pattern:Elbow
- Désignation:Fitting, PVDF, Elbow, Hose Barb Union, 3/32" ID; 10/PK
- Environmentally Preferable:
- Orifice dimensions:0,0625"
- Sterilisation:Autoclave or ETO
- Cdt:10
- Fitting material:PVDF
Frequently Bought Together