Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 5/16" ID x 1/4" ID; 10/PK
N° de catalogue MFLX40614-62
Fournisseur: Avantor Fluid Handling
À propos de cet article
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range −80 to 265 °F (−62 to 129 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Taille de l'ardillon/Tube Ø int. (port 1):⁵⁄₁₆"
- Taille de l'ardillon/Tube Ø int. (port 2):¼"
- Type de connection:Barbed Reducer
- Flow pattern:Straight
- Désignation:Fitting, PVDF, Straight, Hose Barb Reducer, ⁵⁄₁₆" ID x ¼" ID; 10/PK
- Environmentally Preferable:
- Orifice dimensions:3,175 mm (0,125")
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Cdt:10
- Fitting material:PVDF
Frequently Bought Together