Microscope cover glasses, DIN ISO 8255 compliant, 12x24 mm, borosilicate glass D 263 M, thickness No. 1.5H (0.17±0.005 mm), CE marked, 100 pcs./hinged-lid plastic box 1 * 1.000 Pce
N° de catalogue MARI0107342
Fournisseur: MARIENFELD
Frequently Bought Together