Tank 100L stainless steel 304, Single jacket, not suitable for heating/cooling, No lid, no skirts, Suitable for Quattromixer system, Equipped with cleanroom wheels 1 * 1 Pce
N° de catalogue JMBVAFH0100LSSOOQM
Fournisseur: JM SEPARATIONS
Frequently Bought Together