Masterflex® Fitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
Catalog # MFLX40619-99
Supplier: Avantor Fluid Handling
About this item
Build leakproof fluid handling systems.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
These one-way stopcocks feature an all-black PVDF construction.
- Flow pattern:Three-Way
- Désignation:Fitting, PVDF Body with Polypropylene Insert, Female Luer Lock x Three-Way Stopcock
- Seq:06
- Environmentally Preferable:
- Wetted materials:Natural PVDF, Polypropylene
- Cdt:1
- Inlet port:Female Luer Lock
- Outlet port:Female Luer Lock
Frequently Bought Together