Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Low-Profile Hose Barb Adapter, 11/64" ID; 25/PK
N° de catalogue MFLX45518-13
Fournisseur: Avantor Fluid Handling
CE certificate
À propos de cet article
Easily transition to a luer fitting.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Male Luer Lock to Low-Profile Hose Barb, 11/64" ID
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilisation.
- Taille de l'ardillon/Tube Ø int. (port 1):11/64"
- Flow pattern:Straight
- Désignation:Luer Adapters, Male Luer Lock to 11/64" ID low profile barb, PP, 25/Pk
- Seq:14
- Environmentally Preferable:
- Modèle:45518-13
- Connection type (port 1):Male luer lock
- Connection type (port 2):Hose barb
- No. of ports:2
- Cdt:25
- Fitting material:Polypropylene
Frequently Bought Together