Masterflex® Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/8" ID x 10-32 UNF(M); 100/PK
N° de catalogue MFLX40625-06
Fournisseur: Avantor Fluid Handling
À propos de cet article
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/8" ID x 10-32 UNF(M)
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Taille de l'ardillon/Tube Ø int. (port 2):1/8"
- Type de connection:Thread to Barb
- Flow pattern:Straight
- Désignation:Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/8" ID x 10-32 UNF(M); 100/PK
- Seq:18
- Environmentally Preferable:
- Modèle:40625-06
- Sterilisation:Ethylene Oxide (ETO)
- Cdt:100
- Fitting material:Nylon
- Thread size UNF(M) (port 2):10-32"
Frequently Bought Together