Masterflex® Fitting, 316 Stainless Steel, Elbow, Hosebarb to Thread, 1/8" ID x 10-32 UNF(M)
Catalog # MFLX31209-46
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/8" ID x 10-32 UNF(M)
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Taille de l'ardillon/Tube Ø int. (port 2):1/8"
- Type de connection:Thread to Barb
- Flow pattern:Elbow
- Désignation:Fitting, 316 Stainless Steel, Elbow, Hosebarb to Thread, 1/8" ID x 10-32 UNF(M)
- Seq:17
- Environmentally Preferable:
- Modèle:31209-46
- Orifice dimensions:0,09375"
- Garantie:1 year
- Cdt:1
- Fitting material:Stainless Steel 316
- Thread size UNF(M) (port 2):10-32"
Frequently Bought Together