Masterflex® Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 1" ID x 1" ID x 3/4" NPT(M); 100/PK
N° de catalogue MFLX30627-84
Fournisseur: Avantor Fluid Handling
CE certificate
À propos de cet article
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1" ID x 1" ID x 3/4" NPT(M)
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Taille de l'ardillon/Tube Ø int. (port 2):1"
- Type de connection:Thread to Barb
- Flow pattern:T
- Désignation:Fitting, HDPE, Tee, Hosebarb to Thread Adapter, 1" ID x 1" ID x 3/4" NPT(M); 100/PK
- Seq:16
- Environmentally Preferable:
- Modèle:30627-84
- Orifice dimensions:0,78125"
- Sterilisation:Ethylene oxide (ETO) only
- Cdt:100
- Fitting material:HDPE
Frequently Bought Together