Masterflex® Replacement rotor assembly for L/S Standard long-shaft pump heads: 0701X-10, -50
Catalog # MFLX07013-76
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Désignation:Replacement rotor assembly for L/S Standard long-shaft pump heads: 0701X-10, -50
- Seq:01
- Environmentally Preferable:
- Modèle:07013-76
- Cdt:1
- Rotor material:Cold-rolled steel (CRS)
Frequently Bought Together