Masterflex® Syringe Pump Daisy Chain Cable for 74901-80 Pump
N° de catalogue MFLX74901-87
Fournisseur: Avantor Fluid Handling
Spécifications
Voir toutes les configurations du produit- Catalog No:
- MFLX74901-87
- MFLX74901-88
- Modèle:
- 74901-87
- 74901-88
- Outlet connection:
- Daisy Chain Cable for 74901-80
- RS-232 Connector Cable for 74901-80
- Garantie:
- 1 year
- 1 year
- Affichage actuel
Spécifications
À propos de cet article
Daisy chain up to 100 pumps.
- Connect multiple independent syringes to operate simultaneously
- Easily connect pumps
- Choose cable based on your current pump
- For use with 74901-80 Dual Drive Syringe Pump
Optimize performance of your Masterflex® Dual-Drive Syringe Pump with these accessories.
- Désignation:Pumps, Syringe Pumps Daisy Chain Cable for 74901-80 Dual Drive Syringe Pump
- Seq:01
- Environmentally Preferable:
- Modèle:74901-87
- Outlet connection:Daisy Chain Cable for 74901-80
- Garantie:1 year
- Cdt:1
Frequently Bought Together