Masterflex® Fitting, Brass, Straight, Hosebarb to Thread Adapter, 3/8" ID x 3/4" NPT(M); 5/PK
N° de catalogue MFLX30904-53
Fournisseur: Avantor Fluid Handling
À propos de cet article
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Taille de l'ardillon/Tube Ø int. (port 2):⅜"
- Type de connection:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):¾"
- Désignation:Fitting, Brass, Straight, Hosebarb to Thread Adapter, ⅜" ID × ¾" NPT(M); 5/PK
- Seq:15
- Environmentally Preferable:
- Modèle:30904-53
- No. of ports:2
- Garantie:1 year
- Cdt:5
- Fitting material:Brass
Frequently Bought Together