Masterflex® I/P® Easy-Load® Rotor and Bearing Support Assembly
Catalog # MFLX77601-68
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
- Suitable for use with I/P® Easy-Load® pump heads
Replacement parts to keep your pump head operating even longer.
- Pump series:I/P
- Désignation:I/P Rotor/Bearing Support Assembly for Easy-Load 77601-60
- Environmentally Preferable:
- Cdt:1
- Rotor material:Polyphenylene Sulfide (PPS)
Frequently Bought Together