Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for Precision Tubing, SS
N° de catalogue MFLX77200-68
Fournisseur: Avantor Fluid Handling
À propos de cet article
Get the most our of your Masterflex® L/S® Easy-Load® II Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® pump head
Replace your pump head rotor assembly to extend the life of your pump head.
- Pump series:L/S
- Désignation:Replacement rotor assembly for L/S Easy-Load II pump heads 77200-60,
- Environmentally Preferable:
- Garantie:2 years
- Cdt:1
- Rotor material:Stainless Steel
Frequently Bought Together