Masterflex® Fitting, Polypropylene, Elbow, Hose Barb Union, 3/32" ID; 10/PK
N° de catalogue MFLX50622-64
Fournisseur: Avantor Fluid Handling
CE certificate
À propos de cet article
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/32" ID
Create 90° bends in flexible tubing—without the kinks!
- Taille de l'ardillon/Tube Ø int. (port 1):³⁄₃₂"
- Taille de l'ardillon/Tube Ø int. (port 2):³⁄₃₂"
- Type de connection:Barbed Union
- Flow pattern:Elbow
- Désignation:Fitting, Polypropylene, Elbow, Hose Barb Union, ³⁄₃₂" ID; 10/PK
- Seq:29
- Environmentally Preferable:
- Modèle:50622-64
- Sterilisation:Ethylene Oxide (ETO) or Autoclave
- Cdt:10
- Fitting material:Polypropylene
Frequently Bought Together