Masterflex® Flowmeter and Flow Sensor Accessories, Avantor®
Masterflex Ultrasonic flow sensor communications package, 33501-13, 33501-14, 33501-15, 33501-16, 33501-17, 33501-18, 33501-24, 33501-25, 33501-26, 33501-35, 33501-36, 33501-73
N° de catalogue MFLX33501-96
Fournisseur: Avantor Fluid Handling
Masterflex® Flowmeter and Flow Sensor Accessories, Avantor®
N° de catalogue MFLX33501-96
Fournisseur: Avantor Fluid Handling
Aller à :
Product Details & Documents
Make the most of your Masterflex flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
These power supplies, adapters, and other accessories are suitable for use with your Masterflex flowmeter and flow sensors.
Spécifications
- Désignation:Masterflex Ultrasonic flow sensor communications package
- Seq:07
- Compatible avec:33501-13, 33501-14, 33501-15, 33501-16, 33501-17, 33501-18, 33501-24, 33501-25, 33501-26, 33501-35, 33501-36, 33501-73
- Cat. No.:MFLX33501-96
- Connexions:One USB connects to computor for power and data, 2nd USB for addtional power, M12 attaches to Masterflex Flow Sensor
- Garantie:1 year
- Modèle:33501-96