Masterflex® Fitting, Clear Polypropylene, Straight, Hose Barb to Threaded Adapter, 5/32" ID x 10-32 UNF(M); 25/PK
Catalog # MFLX31501-53
Supplier: Avantor Fluid Handling
About this item
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Taille de l'ardillon/Tube Ø int. (port 2):5/32"
- Type de connection:Thread to Barb
- Flow pattern:Straight
- Désignation:Fitting, Clear Polypropylene, Straight, Hose Barb to Threaded Adapter, 5/32" ID x 10-32 UNF(M); 25/PK
- Seq:07
- Environmentally Preferable:
- Modèle:31501-53
- Orifice dimensions:0,7925 mm (0,03125")
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Cdt:25
- Fitting material:Polypropylene
- Thread size UNF(M) (port 2):10-32"
Frequently Bought Together