Masterflex® Fitting, Nylon, Elbow, Hosebarb to Thread Adapter, 1/4" ID x 1/8" NPT(M); 100/PK
N° de catalogue MFLX30401-07
Fournisseur: Avantor Fluid Handling
À propos de cet article
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/4" ID x 1/8" NPT(M)
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Taille de l'ardillon/Tube Ø int. (port 2):1/4"
- Type de connection:Thread to Barb
- Flow pattern:Elbow
- Thread size NPT(M) (port 1):1/8"
- Désignation:Fitting, Nylon, Elbow, Hosebarb to Thread Adapter, 1/4" ID x 1/8" NPT(M); 100/PK
- Seq:03
- Environmentally Preferable:
- Modèle:30401-07
- Orifice dimensions:0,15625"
- Sterilisation:Ethylene oxide (ETO) only
- Garantie:1 year
- Cdt:100
- Fitting material:Nylon
Frequently Bought Together