ContactUsLinkComponent
Masterflex® L/S® Replacement Gear Assembly for 6 to 600 rpm Drives
Masterflex® L/S® Replacement Gear Assembly for 6 to 600 rpm Drives
Catalog # MFLX07553-09
undefined
Masterflex® L/S® Replacement Gear Assembly for 6 to 600 rpm Drives
Catalog # MFLX07553-09
About this item
Make the most of your Masterflex drives replacement parts and service kits.
  • Easy to install, these service kits avoid costly downtime of your gear pump head
Extend the life of your Masterflex pump with these replacement parts.
  • Désignation:
    Replacement gear assembly for L/S 6-600 rpm drives.
  • Environmentally Preferable:
  • Compatible avec:
    07520-00, -20, -25, -40, -45, 07521-00, -20, -25, -40, -45, 07522-10, -15, 07523-00, -02, -20, -25, 07525-00, -02, -34, -36, 07526-00, -05, 07527-00,
  • Cdt:
    1
Frequently Bought Together