Masterflex® L/S® Easy-Load® Pump Head Replacement Open-Head Sensor for 07514-series
Catalog # MFLX07514-08
Supplier: Avantor Fluid Handling
1 year warranty
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Désignation:L/S® Easy-Load® Pump Head Replacement Open-Head Sensor for 07514-series
- Seq:35
- Environmentally Preferable:
- Modèle:07514-08
- Open-head sensor:Yes
- Garantie:1 year
- Cdt:1
Frequently Bought Together