Masterflex® I/P® Standard Pump Head, Replacement Thrust Washers; 10/Pk
Catalog # MFLX07019-92
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with I/P® Standard pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- Désignation:Standard Pump Head, Replacement Thrust Washers; 10/Pk
- Seq:29
- Environmentally Preferable:
- Modèle:07019-92
- Open-head sensor:No
- Tubing sizes accepted:I/P
- Cdt:10
Frequently Bought Together