Masterflex® I/P® Replacement Rotor Assembly, Long-Shaft, CRS
Catalog # MFLX07019-80
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- Désignation:Replacement Rotor Assembly, Long-Shaft, CRS
- Seq:25
- Environmentally Preferable:
- Modèle:07019-80
- Open-head sensor:No
- Garantie:2 years
- Cdt:1
- Rotor material:Cold-rolled steel (CRS)
Frequently Bought Together