Masterflex® I/P® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, I/P® 73
N° de catalogue MFLX07019-78
Fournisseur: Avantor Fluid Handling
À propos de cet article
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for use with I/P® 73 Standard pump head
- Constructed with durable polycarbonate (PC) housing and cold-rolled steel (CRS) rotor
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- Désignation:Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, CRS Rotor, I/P® 73 Tubing
- Seq:23
- Environmentally Preferable:
- Modèle:07019-78
- Open-head sensor:No
- Tubing sizes accepted:I/P 73
- Garantie:2 years
- Cdt:1
- Matériau du boîtier:PC
- Rotor material:Cold-rolled steel (CRS)
Frequently Bought Together