Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Pump head mounting hardware, Standard and Easy-Load®, four pump heads, stainless steel; 2/set, 2 years warranty, Acier inoxydable, Masterflex L/S® Easy-Load® Pump Head, L/S
N° de catalogue MFLX07013-09
Fournisseur: Avantor Fluid Handling
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
N° de catalogue MFLX07013-09
Fournisseur: Avantor Fluid Handling
Aller à :
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Désignation:L/S® Pump head mounting hardware, Standard and Easy-Load®, four pump heads, stainless steel; 2/set, 2 years warranty
- Matière:Acier inoxydable
- Seq:04
- Compatible avec:Masterflex L/S® Easy-Load® Pump Head
- Pump series:L/S