Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Pump head mounting hardware, Standard and Easy-Load®, one Pump Head, Stainless Steel; 2/Set, 2 years warranty, Acier inoxydable, Masterflex L/S® Easy-Load® Pump Head, L/S
N° de catalogue MFLX07013-04
Fournisseur:   Avantor Fluid Handling
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
N° de catalogue MFLX07013-04
Fournisseur:  Avantor Fluid Handling
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Pump head mounting hardware, Standard and Easy-Load®, one Pump Head, Stainless Steel; 2/Set, 2 years warranty, Acier inoxydable, Masterflex L/S® Easy-Load® Pump Head, L/S
Prix
67,50 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
  • Constructed of durable stainless steel
  • See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Spécifications
  • Désignation:
    L/S® Pump head mounting hardware, Standard and Easy-Load®, one Pump Head, Stainless Steel; 2/Set, 2 years warranty
  • Matière:
    Acier inoxydable
  • Seq:
    01
  • Compatible avec:
    Masterflex L/S® Easy-Load® Pump Head
  • Pump series:
    L/S