Fitting, Polypropylene, Elbow, Hose Barb Union, 5/32" ID; 10/PK
N° de catalogue MFLX41518-58
Fournisseur: Avantor Fluid Handling
À propos de cet article
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- Taille de l'ardillon/Tube Ø int. (port 1):⁵⁄₃₂"
- Taille de l'ardillon/Tube Ø int. (port 2):⁵⁄₃₂"
- Type de connection:Barbed Union
- Flow pattern:Straight
- Désignation:Fitting, Polypropylene, Elbow, Hose Barb Union, ⁵⁄₃₂" ID; 10/PK
- Seq:27
- Environmentally Preferable:
- Modèle:41518-58
- Cdt:10
- Fitting material:Polypropylene
Frequently Bought Together