Masterflex® Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16" ID x 1/8" NPT(M); 100/PK
N° de catalogue MFLX40626-86
Fournisseur: Avantor Fluid Handling
CE certificate
À propos de cet article
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Fitting, Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/4" ID x 3/4" ID x 3/4" NPT(M); 100/PK
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Taille de l'ardillon/Tube Ø int. (port 2):5/16"
- Type de connection:Thread to Barb
- Flow pattern:Elbow
- Thread size NPT(M) (port 1):1/8"
- Désignation:Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16" ID x 1/8" NPT(M); 100/PK
- Seq:22
- Environmentally Preferable:
- Modèle:40626-86
- Orifice dimensions:0,1875"
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Cdt:100
- Fitting material:Polypropylene
Frequently Bought Together