Masterflex® Fitting, Polypropylene, Elbow, Hosebarb to Thread, 3/16" ID x 1/4" NPT(M); 10/PK
N° de catalogue MFLX40622-95
Fournisseur: Avantor Fluid Handling
CE certificate
À propos de cet article
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/16" ID x 1/4" NPT(M)
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Taille de l'ardillon/Tube Ø int. (port 2):3/16"
- Type de connection:Thread to Barb
- Flow pattern:Elbow
- Thread size NPT(M) (port 1):1/4"
- Désignation:Fitting, Polypropylene, Elbow, Hosebarb to Thread, 3/16" ID x 1/4" NPT(M); 10/PK
- Seq:21
- Environmentally Preferable:
- Modèle:40622-95
- Orifice dimensions:0,09375"
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Cdt:10
- Fitting material:Polypropylene
Frequently Bought Together